[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: ((Tetrahydro-2H-pyran-4-yl)methyl)hydrazine dihydrochloride

OR306224-250MG
APOSOR306224-250MGEA 238 EUR
APOSOR306224-250MG
[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride 95+%
[(Tetrahydro-2H-pyran-4-yl)methyl]hydrazine dihydrochloride
Formel: C₆H₁₄N₂O·2ClH
Molecular Weight: 203,11 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19382069
CAS-Nummer: 1315365-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR