3,4,5-Trihydroxybenzaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: Gallic aldehyde, Pyrogallol-5-carboxaldehyde

OR30631-25G OR30631-5G
APOSOR30631-25GEA 412 EUR
APOSOR30631-25G APOSOR30631-5G
3,4,5-Trihydroxybenzaldehyd
3,4,5-Trihydroxybenzaldehyde
Formel: C₇H₆O₄
Molecular Weight: 154.12 g/mol
Schmelzpunkt: 211…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003371
CAS-Nummer: 13677-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR