(4-Methoxypyridin-2-yl)methanamine 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Aminomethyl)-4-methoxypyridine

OR306389-1G
APOSOR306389-1GEA 94.4 EUR
APOSOR306389-1G
(4-Methoxypyridin-2-yl)methanamine 98%
(4-Methoxypyridin-2-yl)methanamine
Formel: C₇H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD11111886
CAS-Nummer: 194658-14-5
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR