3-Benzyl-3,9-diazaspiro[5.5]undecane 95+%

Lieferant: Apollo Scientific

OR306396-250MG
APOSOR306396-250MGEA 39.3 EUR
APOSOR306396-250MG
3-Benzyl-3,9-diazaspiro[5.5]undecane 95+%
3-Benzyl-3,9-diazaspiro[5.5]undecane
Formel: C₁₆H₂₄N₂
Molecular Weight: 244,375 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035940
CAS-Nummer: 189333-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR