2-(Aminomethyl)-1,3-oxazole-4-carboxylic acid, 2-BOC protected 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-{[(tert-Butoxycarbonyl)amino]methyl}-1,3-oxazole-4-carboxylic acid
OR30642-100MG
APOSOR30642-100MGEA
127
EUR
APOSOR30642-100MG
2-(Aminomethyl)-1,3-oxazole-4-carboxylic acid, 2-BOC protected 95%
2-(Aminomethyl)-1,3-oxazole-4-carboxylic acid, 2-BOC protected
Formel:
C₁₀H₁₄N₂O₅ Molecular Weight: 242,229 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08236858 CAS-Nummer: 182120-90-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...