4-(4-Bromophenyl)oxane-4-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-(4-Bromophenyl)tetrahydro-2H-pyran-4-carboxylic acid

OR306432-250MG
APOSOR306432-250MGEA 173 EUR
APOSOR306432-250MG
4-(4-Bromophenyl)oxane-4-carboxylic acid 95+%
4-(4-Bromophenyl)oxane-4-carboxylic acid
Formel: C₁₂H₁₃BrO₃
Molecular Weight: 285,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10693768
CAS-Nummer: 1152567-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR