2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
OR30644-50MG
APOSOR30644-50MGEA
0
EUR
APOSOR30644-50MG
2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
2-(Aminomethyl)-1H-imidazole-5-carboxylic acid, 2-BOC protected
Formel:
C₁₀H₁₅N₃O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10703328 CAS-Nummer: 867340-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...