(5-Methylpyridin-2-yl)methanamine
Lieferant: Apollo Scientific
|
Gefahr
|
OR306469-1G
OR306469-500MG
OR306469-5G
APOSOR306469-1GEA
172
EUR
APOSOR306469-1G
APOSOR306469-500MG
APOSOR306469-5G
(5-Methylpyridin-2-yl)methanamine
(5-Methylpyridin-2-yl)methanamine
Formel:
C₇H₁₀N₂ Siedepunkt: 102…105 °C/12mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD08752541 CAS-Nummer: 45715-08-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...