2,8-Diazaspiro[4.5]decan-3-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Oxo-2,8-diazaspiro[4.5]decane hydrochloride
OR306483-250MG
APOSOR306483-250MGEA
21.9
EUR
APOSOR306483-250MG
2,8-Diazaspiro[4.5]decan-3-one hydrochloride
2,8-Diazaspiro[4.5]decan-3-one hydrochloride
Formel:
C₈H₁₄N₂O·ClH Molecular Weight: 190,671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520496 CAS-Nummer: 945892-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...