4-Methoxy-1H-pyrrolo[2,3-b]pyridine 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-7-azaindole

OR30653-1G
APOSOR30653-1GEA 55.1 EUR
APOSOR30653-1G
4-Methoxy-1H-pyrrolo[2,3-b]pyridine 95+%
4-Methoxy-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272230
CAS-Nummer: 122379-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR