6-Bromo-2-methylisoindolin-1-one

Lieferant: Apollo Scientific
Achtung

OR306541-1G
APOSOR306541-1GEA 220 EUR
APOSOR306541-1G
6-Bromo-2-methylisoindolin-1-one
6-Bromo-2-methylisoindolin-1-one
Formel: C₉H₈BrNO
Molecular Weight: 226,07 g/mol
Schmelzpunkt: 114…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17676936
CAS-Nummer: 1254319-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR