5-Cyclopropylpicolinic acid

Lieferant: Apollo Scientific

OR306657-1G OR306657-250MG
APOSOR306657-1GEA 574 EUR
APOSOR306657-1G APOSOR306657-250MG
5-Cyclopropylpicolinic acid
5-Cyclopropylpicolinic acid
Formel: C₉H₉NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD17259279
CAS-Nummer: 1174322-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR