3-Cyclopropylisonicotinic acid
Lieferant: Apollo Scientific
Synonyme:
3-Cyclopropylpyridine-4-carboxylic acid
OR306658-1G
OR306658-250MG
APOSOR306658-1GEA
575
EUR
APOSOR306658-1G
APOSOR306658-250MG
3-Cyclopropylisonicotinic acid
3-Cyclopropylisonicotinic acid
Formel:
C₉H₉NO₂ Molecular Weight: 163.173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18261166 CAS-Nummer: 1256785-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...