2-Cyclopropylisonicotinic acid

Lieferant: Apollo Scientific
Achtung

OR306659-1G OR306659-250MG
APOSOR306659-1GEA 429 EUR
APOSOR306659-1G APOSOR306659-250MG
2-Cyclopropylisonicotinic acid
2-Cyclopropylisonicotinic acid
Formel: C₉H₉NO₂
Schmelzpunkt: 285…287 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13187883
CAS-Nummer: 1216171-07-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR