5-Brom-2-methoxy-nicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

OR30675-10G OR30675-25G
APOSOR30675-10GEA 85.7 EUR
APOSOR30675-10G APOSOR30675-25G
5-Brom-2-methoxy-nicotinsäure 97%
5-Brom-2-methoxy-nicotinsäure
Formel: C₇H₆BrNO₃
Molecular Weight: 232.04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656771
CAS-Nummer: 54916-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR