5-Brom-2-methoxy-nicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR30675-10G
OR30675-25G
APOSOR30675-10GEA
85.7
EUR
APOSOR30675-10G
APOSOR30675-25G
5-Brom-2-methoxy-nicotinsäure 97%
5-Brom-2-methoxy-nicotinsäure
Formel:
C₇H₆BrNO₃ Molecular Weight: 232.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656771 CAS-Nummer: 54916-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...