(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: trans-4-Amino-L-proline dihydrochloride

OR30676-1G
APOSOR30676-1GEA 635 EUR
APOSOR30676-1G
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride
Formel: C₅H₁₀N₂O₂·2ClH
Molecular Weight: 203,067 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954286
CAS-Nummer: 16257-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR