1-tert-Butyl 2-methyl (2R,4S)-4-aminopyrrolidine-1,2-dicarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,4S)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate, Methyl (2R,4S)-4-aminopyrrolidine-2-carboxylate, N1-BOC protected, trans-4-Amino-D-proline methyl ester, N1-BOC protected, (2R,4S)-4-amino-1-(tert-butoxycarbonyl)-2-(methoxycarbonyl)pyrrolidine

OR30679-250MG
APOSOR30679-250MGEA 43.5 EUR
APOSOR30679-250MG
1-tert-Butyl 2-methyl (2R,4S)-4-aminopyrrolidine-1,2-dicarboxylate
1-tert-Butyl 2-methyl (2R,4S)-4-aminopyrrolidine-1,2-dicarboxylate
Formel: C₁₁H₂₀N₂O₄
Molecular Weight: 244.288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08704545
CAS-Nummer: 254881-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR