Bis(triphenylphosphin)palladium(II)chlorid 98%

Lieferant: Apollo Scientific
Achtung

OR30692-1G OR30692-25G OR30692-5G
APOSOR30692-1GEA 40.6 EUR
APOSOR30692-1G APOSOR30692-25G APOSOR30692-5G
Bis(triphenylphosphin)palladium(II)chlorid 98%
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Lagertemperatur: Kühlschrank
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR