1H-Pyrazolo[3,4-c]pyridine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR30697-1G
APOSOR30697-1GEA
23.2
EUR
APOSOR30697-1G
1H-Pyrazolo[3,4-c]pyridine 95+%
1H-Pyrazolo[3,4-c]pyridine
Formel:
C₆H₅N₃ Molecular Weight: 119,1240 g/mol Schmelzpunkt: 100…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661308 CAS-Nummer: 271-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...