4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%

Lieferant: Apollo Scientific

OR307014-1G
APOSOR307014-1GEA 194 EUR
APOSOR307014-1G
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine 98%
4,6-Dichloro-2-(methylthio)-5-nitropyrimidine
Formel: C₅H₃Cl₂N₃O₂S
Molecular Weight: 240,0672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02323208
CAS-Nummer: 1979-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR