(R,R)-Cyclohex-4-ene-1,2-dicarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR307025-1G
APOSOR307025-1GEA 137 EUR
APOSOR307025-1G
(R,R)-Cyclohex-4-ene-1,2-dicarboxylic acid
(R,R)-Cyclohex-4-ene-1,2-dicarboxylic acid
Formel: C₈H₁₀O₄
Molecular Weight: 170,163 g/mol
Schmelzpunkt: 167…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09757670
CAS-Nummer: 15573-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR