(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1S,2R)-1-Methyl cis-4-cyclohexene-1,2-dicarboxylate, (1S,2R)-1,2,3,6-Tetrahydrophthalic acid 2-hydrogen 1-methyl ester, (1S,2R)-1-Methyl cis-1,2,3,6-tetrahydrophthalate, 2-Hydrogen 1-Methyl (1S,2R)-1,2,3,6-tetrahydrophthalate, (1R,6S)-cis-6-(Methoxycarbonyl)cyclohex-3-ene-1-carboxylic acid, (1R,6S)-6-(Methoxycarbonyl)cyclohex-3-enecarboxylic acid
OR307028-1G
APOSOR307028-1GEA
116
EUR
APOSOR307028-1G
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester
Formel:
C₉H₁₂O₄ Molecular Weight: 184.19 g/mol Schmelzpunkt: 65…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075490 CAS-Nummer: 88335-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...