1-(Pyrimidin-5-yl)ethanone 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Acetylpyrimidine
OR30703-250MG
APOSOR30703-250MGEA
29
EUR
APOSOR30703-250MG
1-(Pyrimidin-5-yl)ethanone 98%
1-(Pyrimidin-5-yl)ethanone
Formel:
C₆H₆N₂O Molecular Weight: 122,1246 g/mol Siedepunkt: 70 °C/2mmHg Schmelzpunkt: 87…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09038477 CAS-Nummer: 10325-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...