(R)-1-Cbz-3-aminopiperidine 97%

Lieferant: Apollo Scientific

Synonyme: (R)-Benzyl 3-aminopiperidine-1-carboxylate

OR307040-1G
APOSOR307040-1GEA 27.7 EUR
APOSOR307040-1G
(R)-1-Cbz-3-aminopiperidine 97%
(R)-1-Cbz-3-aminopiperidine
Formel: C₁₃H₁₈N₂O₂
Molecular Weight: 234,294 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864625
CAS-Nummer: 1044560-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR