(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine

OR307060-5G
APOSOR307060-5GEA 36.4 EUR
APOSOR307060-5G
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 96%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
Formel: C₈H₁₆N₂O₄
Molecular Weight: 204.23 g/mol
Schmelzpunkt: 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00236843
CAS-Nummer: 73259-81-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR