trans-N-Boc-4-cyano-L-proline methyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-tert-Butyl 2-methyl (2S,4R)-4-cyanopyrrolidine-1,2-dicarboxylate
OR307067-1G
APOSOR307067-1GEA
812
EUR
APOSOR307067-1G
trans-N-Boc-4-cyano-L-proline methyl ester
trans-N-Boc-4-cyano-L-proline methyl ester
Formel:
C₁₂H₁₈N₂O₄ Molecular Weight: 254.286 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094915 CAS-Nummer: 194163-91-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...