3-Brom-2-nitroanisol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3-methoxy-2-nitrobenzene
OR30707-1G
APOSOR30707-1GEA
21.9
EUR
APOSOR30707-1G
3-Brom-2-nitroanisol 98%
3-Brom-2-nitroanisol
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,0314 g/mol Siedepunkt: 130…131 °C/3mmHg Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101044 CAS-Nummer: 500298-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...