cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR307073-1G
APOSOR307073-1GEA
21.9
EUR
APOSOR307073-1G
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95+%
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline
Formel:
C₁₀H₁₇NO₅ Schmelzpunkt: 146…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02094407 CAS-Nummer: 135042-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...