Boc-D-Pro-OH 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline

OR307074-500G
APOSOR307074-500GEA 165 EUR
APOSOR307074-500G
Boc-D-Pro-OH 97%
Boc-D-Pro-OH
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215.25 g/mol
Schmelzpunkt: 134…137 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063226
CAS-Nummer: 37784-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR