4-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)benzoesäure 98%
Lieferant: Apollo Scientific
Synonyme:
Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid
OR307084-100G
APOSOR307084-100GEA
87
EUR
APOSOR307084-100G
4-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)benzoesäure 98%
4-((tert-Butoxycarbonyl)amino)benzoic acid
Formel:
C₁₂H₁₅NO₄ Molecular Weight: 237,3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037428 CAS-Nummer: 66493-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...