Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-D-Val(3-OH)-OH, (R)-2-((tert-Butoxycarbonyl)amino)-3-hydroxy-3-methylbutanoic acid, 3-Hydroxy-D-valine, N-BOC protected

OR307093-1G
APOSOR307093-1GEA 128 EUR
APOSOR307093-1G
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Boc-(R)-2-amino-3-hydroxy-3-methylbutanoic acid
Formel: C₁₀H₁₉NO₅
Molecular Weight: 233,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682576
CAS-Nummer: 288159-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR