(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%

Lieferant: Apollo Scientific

Synonyme: Boc-Ser(Bzl)-OH, N-Boc-O-benzyl-L-serine

OR307098-100G
APOSOR307098-100GEA 115 EUR
APOSOR307098-100G
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid
Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066063
CAS-Nummer: 23680-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR