(1S,4R)-4-Aminocyclopent-2-ene carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1R,4S)-rel-4-Aminocyclopent-2-enecarboxylic acid

OR307108-1G
APOSOR307108-1GEA 323 EUR
APOSOR307108-1G
(1S,4R)-4-Aminocyclopent-2-ene carboxylic acid
(1S,4R)-4-Aminocyclopent-2-ene carboxylic acid
Formel: C₆H₉NO₂
Molecular Weight: 127,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00211286
CAS-Nummer: 168471-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR