cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: cis-4-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, cis-4-(BOC-amino)cyclohexanecarboxylic acid, cis-4-(Boc-amino)cyclohexanecarboxylic Acid

OR307110-10G
APOSOR307110-10GEA 339 EUR
APOSOR307110-10G
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
cis-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Formel: C₁₂H₂₁NO₄
Molecular Weight: 243,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862294
CAS-Nummer: 53292-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR