N-Boc-(S)-2-amino-3-methoxy-1-propanol

Lieferant: Apollo Scientific
Achtung

OR307124-1G
APOSOR307124-1GEA 372 EUR
APOSOR307124-1G
N-Boc-(S)-2-amino-3-methoxy-1-propanol
N-Boc-(S)-2-amino-3-methoxy-1-propanol
Formel: C₉H₁₉NO₄
Molecular Weight: 205,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20261175
CAS-Nummer: 721927-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR