1-Methyl-1H-indazole-6-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

OR30713-1G
APOSOR30713-1GEA 144 EUR
APOSOR30713-1G
1-Methyl-1H-indazole-6-carboxaldehyde
1-Methyl-1H-indazole-6-carboxaldehyde
Formel: C₉H₈N₂O
Lagertemperatur: Kühlschrank
MDL: MFCD11109366
CAS-Nummer: 1092351-51-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR