(R)-1-(Boc-amino)-2-propanol 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-tert-Butyl (2-hydroxypropyl)carbamate

OR307131-100G OR307131-25G OR307131-5G
APOSOR307131-100GEA 441 EUR
APOSOR307131-100G APOSOR307131-25G APOSOR307131-5G
(R)-1-(Boc-amino)-2-propanol 96%
(R)-1-(Boc-amino)-2-propanol
Formel: C₈H₁₇NO₃
Molecular Weight: 175.23 g/mol
Dichte: 1.02 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD04974338
CAS-Nummer: 119768-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR