Methyl-1H-indazol-5-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 1H-indazole-5-carboxylate
OR30715-1G
APOSOR30715-1GEA
21.9
EUR
APOSOR30715-1G
Methyl-1H-indazol-5-carboxylat
Methyl-1H-indazol-5-carboxylat
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,1700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07371609 CAS-Nummer: 473416-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...