(R)-(-)-1,2-Propandiol 98%

Lieferant: Apollo Scientific

OR307150-500G
APOSOR307150-500GEA 132 EUR
APOSOR307150-500G
(R)-(-)-1,2-Propandiol 98%
(R)-(-)-1,2-Propandiol
Formel: C₃H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00066248
CAS-Nummer: 4254-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR