1-(1-Ethoxyethyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR307169-1G
APOSOR307169-1GEA
58
EUR
APOSOR307169-1G
1-(1-Ethoxyethyl)-1H-pyrazole
1-(1-Ethoxyethyl)-1H-pyrazole
Formel:
C₇H₁₂N₂O Molecular Weight: 140,183 g/mol Flammpunkt: >66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD17676680 CAS-Nummer: 28791-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...