Benzofuran-7-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Formylbenzo[b]furan, 1-Benzofuran-7-carboxaldehyde, Benzo[b]furan-7-carboxaldehyde

OR307183-1G
APOSOR307183-1GEA 168 EUR
APOSOR307183-1G
Benzofuran-7-carbaldehyde
Benzofuran-7-carbaldehyde
Formel: C₉H₆O₂
Molecular Weight: 146,14 g/mol
Siedepunkt: 103…105/1 mm °C
MDL: MFCD08272125
CAS-Nummer: 95333-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR