2-(Furan-3-yl)acetic acid

Lieferant: Apollo Scientific
Gefahr

OR307188-1G OR307188-250MG OR307188-100MG
APOSOR307188-1GEA 0 EUR
APOSOR307188-1G APOSOR307188-250MG APOSOR307188-100MG
2-(Furan-3-yl)acetic acid
2-(Furan-3-yl)acetic acid
Formel: C₆H₆O₃
Lagertemperatur: Kühlschrank
MDL: MFCD01859888
CAS-Nummer: 123617-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR