1-Brom-3,5-dinitrobenzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dinitrobrombenzol
OR307190-1G
OR307190-25G
OR307190-5G
APOSOR307190-1GEA
21.9
EUR
APOSOR307190-1G
APOSOR307190-25G
APOSOR307190-5G
1-Brom-3,5-dinitrobenzol 99%
1-Bromo-3,5-dinitrobenzene
Formel:
C₆H₃BrN₂O₄ Molecular Weight: 247 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156596 CAS-Nummer: 18242-39-2 UN: 3459 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...