1-Brom-3,5-dinitrobenzol 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dinitrobrombenzol

OR307190-1G OR307190-25G OR307190-5G
APOSOR307190-1GEA 21.9 EUR
APOSOR307190-1G APOSOR307190-25G APOSOR307190-5G
1-Brom-3,5-dinitrobenzol 99%
1-Bromo-3,5-dinitrobenzene
Formel: C₆H₃BrN₂O₄
Molecular Weight: 247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156596
CAS-Nummer: 18242-39-2
UN: 3459
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR