3-(Chloromethyl)-2-hydroxy-5-methylbenzaldehyde 95%

Lieferant: Apollo Scientific
Gefahr

OR307192-1G
APOSOR307192-1GEA 328 EUR
APOSOR307192-1G
3-(Chloromethyl)-2-hydroxy-5-methylbenzaldehyde 95%
3-(Chloromethyl)-2-hydroxy-5-methylbenzaldehyde
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03055723
CAS-Nummer: 192819-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR