(4-Methoxy-3-methylpyridin-2-yl)methanol 97%

Lieferant: Apollo Scientific

OR307212-250MG
APOSOR307212-250MGEA 84.1 EUR
APOSOR307212-250MG
(4-Methoxy-3-methylpyridin-2-yl)methanol 97%
(4-Methoxy-3-methylpyridin-2-yl)methanol
Formel: C₈H₁₁NO₂
Molecular Weight: 153,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272123
CAS-Nummer: 86604-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR