2-Methyl-2H-indazole-4-carboxylic acid methyl ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-methyl-2H-indazole-4-carboxylate
OR30722-250MG
APOSOR30722-250MGEA
24.8
EUR
APOSOR30722-250MG
2-Methyl-2H-indazole-4-carboxylic acid methyl ester
2-Methyl-2H-indazole-4-carboxylic acid methyl ester
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190,2 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11109405 CAS-Nummer: 1071428-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...