4-Methoxy-2,3-dimethylpyridine

Lieferant: Apollo Scientific
Achtung

OR307220-1G
APOSOR307220-1GEA 127 EUR
APOSOR307220-1G
4-Methoxy-2,3-dimethylpyridine
4-Methoxy-2,3-dimethylpyridine
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11100693
CAS-Nummer: 163593-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR