N-(Pyridin-4-yl)pyridine-4-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Pyridin-4-yl)isonicotinamide

OR307226-1G
APOSOR307226-1GEA 92.8 EUR
APOSOR307226-1G
N-(Pyridin-4-yl)pyridine-4-carboxamide
N-(Pyridin-4-yl)pyridine-4-carboxamide
Formel: C₁₁H₉N₃O
Molecular Weight: 199,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00454168
CAS-Nummer: 64479-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR